Thermo Scientific Chemicals

Aluminum bromide, 98%, Thermo Scientific Chemicals

Catalog number: 011113.22
100 g, Each
Thermo Scientific Chemicals

Aluminum bromide, 98%, Thermo Scientific Chemicals

Catalog number: 011113.22
100 g, Each
Quantity
Catalog number: 011113.22
also known as 011113-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7727-15-3
IUPAC Name
aluminium(3+) tribromide
Molecular Formula
AlBr3
InChI Key
PQLAYKMGZDUDLQ-UHFFFAOYSA-K
SMILES
[Al+3].[Br-].[Br-].[Br-]
Assay (unspecified)
97.6% min

Description

Alkylation, bromination and isomerization catalyst in organic synthesis.Aluminum bromide acts as a strong Lewis acid and is used in Lewis acid-promoted reactions such as epoxide ring openings. It is used as a catalyst for Friedel-Crafts alkylation reaction. It also finds application in bromination and isomerization reactions in organic synthesis. It is also used in polymerization reaction to prepare poly(o-xylene)from benzocyclobutene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text