Thermo Scientific Chemicals

Calcium sulfate, anhydrous, Puratronic™, 99.993% (metals basis), Thermo Scientific Chemicals

Catalog number: 011116.09
10 g, Each
Thermo Scientific Chemicals

Calcium sulfate, anhydrous, Puratronic™, 99.993% (metals basis), Thermo Scientific Chemicals

Catalog number: 011116.09
10 g, Each
Quantity
Catalog number: 011116.09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7778-18-9
IUPAC Name
calcium sulfate
Molecular Formula
CaO4S
InChI Key
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES
[Ca++].[O-]S([O-])(=O)=O
Total Metal Impurities
0.007% max.

Description

Calcium sulfate, anhydrous is used as adhesives and sealant chemical, Fillers, Fuels and fuel additives, Intermediates, Process regulators, Agricultural Products, Building/Construction Materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text