Zinc nitrate hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Zinc nitrate hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc nitrate hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 011136.18
also known as 011136-18
Price (USD)/ Each
250.65
Online exclusive
278.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
250.65
Online exclusive
278.00 
Save 27.35 (10%)
Add to cart
Zinc nitrate hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number011136.18
Price (USD)/ Each
250.65
Online exclusive
278.00 
Save 27.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10196-18-6
IUPAC Namezinc(2+) hexahydrate dinitrate
Molecular FormulaH12N2O12Zn
InChI KeyJGPSMWXKRPZZRG-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Zinc nitrate hexahydrate is used as an analytical reagent, catalyst and as a mordant in dyeing. It finds application in electro galvanizing, rubber industry and in the water treatment industry. It is widely used as the precursor for the synthesis of zinc oxide. It is employed in the preparation of the steel phosphating agent. In the field of agriculture, it is used for nourishing and protecting fruit trees from acid rain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only