Thermo Scientific Chemicals

Zinc sulfate hydrate, Puratronic, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 011138.18
50 g, Each
Thermo Scientific Chemicals

Zinc sulfate hydrate, Puratronic, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 011138.18
50 g, Each
Quantity
Catalog number: 011138.18
also known as 011138-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16788-42-4
IUPAC Name
zinc(2+) hydrate sulfate
Molecular Formula
H2O5SZn
InChI Key
RNZCSKGULNFAMC-UHFFFAOYSA-L
SMILES
O.[Zn++].[O-]S([O-])(=O)=O
Form
Powder or crystals or crystalline powder
Total Metal Impurities
≤0.002%

Description

Zinc sulfate hydrate is used in the production of rayon, as a feed supplement, and as a fertilizer ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text