Thermo Scientific Chemicals

Gallium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 011150.14
25 g, Each
Thermo Scientific Chemicals

Gallium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 011150.14
25 g, Each
Quantity
Catalog number: 011150.14
also known as 011150-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69365-72-6
IUPAC Name
gallium(3+) trinitrate
Molecular Formula
GaN3O9
InChI Key
CHPZKNULDCNCBW-UHFFFAOYSA-N
SMILES
[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Metal Impurities
0.001% max.

Description

Gallium(III) nitrate hydrate is the potential precursor for the synthesis of other gallium compounds. It finds application in the preparation of gallium nitride, which is used as a solar cell array for satellites. In the field of pharmaceutical industry, it is used to treat symptomatic hypercalcemia by preventing the breakdown of bone by slowing osteoclast activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text