Gallium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Gallium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gallium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Catalog number 011150.36
also known as 011150-36
Price (USD)/ Each
3,899.65
Online exclusive
4333.00 
Save 433.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
3,899.65
Online exclusive
4333.00 
Save 433.35 (10%)
Add to cart
Gallium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number011150.36
Price (USD)/ Each
3,899.65
Online exclusive
4333.00 
Save 433.35 (10%)
-
Add to cart
Chemical Identifiers
CAS69365-72-6
IUPAC Namegallium(3+) trinitrate
Molecular FormulaGaN3O9
InChI KeyCHPZKNULDCNCBW-UHFFFAOYSA-N
SMILES[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Gallium(III) nitrate hydrate is the potential precursor for the synthesis of other gallium compounds. It finds application in the preparation of gallium nitride, which is used as a solar cell array for satellites. In the field of pharmaceutical industry, it is used to treat symptomatic hypercalcemia by preventing the breakdown of bone by slowing osteoclast activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only