Thermo Scientific Chemicals

Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb content reported, Thermo Scientific Chemicals

Catalog number: 011173.04
2 g, Each
Thermo Scientific Chemicals

Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb content reported, Thermo Scientific Chemicals

Catalog number: 011173.04
2 g, Each
Quantity
Catalog number: 011173.04
also known as 011173-04
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1327-53-3
IUPAC Name
diarsorosooxidane
Molecular Formula
As2O3
InChI Key
IKWTVSLWAPBBKU-UHFFFAOYSA-N
SMILES
O=[As]O[As]=O
Appearance (Color)
White
Form
Powder
Impurity content
Sb content reported
Total metal impurities excluding Sb
0.004% max.

Description

Arsenic trioxide is used as a precursor to forestry products, in colorless glass production, and in electronics. And it is also used as pigments, and preservatives for hides and wood, in the semiconductor industry to create light emitting diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text