Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb content reported, Thermo Scientific Chemicals
Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb content reported, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb content reported, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011173.09
also known as 011173-09
Price (USD)/ Each
68.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
80.10 
Save 11.45 (14%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
68.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
80.10 
Save 11.45 (14%)
Add to cart
Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb content reported, Thermo Scientific Chemicals
Catalog number011173.09
Price (USD)/ Each
68.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
80.10 
Save 11.45 (14%)
-
Add to cart
Chemical Identifiers
CAS1327-53-3
IUPAC Namediarsorosooxidane
Molecular FormulaAs2O3
InChI KeyIKWTVSLWAPBBKU-UHFFFAOYSA-N
SMILESO=[As]O[As]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Impurity contentSb content reported
Total metal impurities excluding Sb0.004% max.
Arsenic trioxide is used as a precursor to forestry products, in colorless glass production, and in electronics. And it is also used as pigments, and preservatives for hides and wood, in the semiconductor industry to create light emitting diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only