Yttrium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Yttrium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 011184.22
also known as 011184-22
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
Add to cart
Yttrium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Catalog number011184.22
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12741-05-8
IUPAC Nameyttrium(3+) trichloride
Molecular FormulaCl3Y
InChI KeyPCMOZDDGXKIOLL-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay from Suppliers CofATotal Rare Earth Oxide Impurities: ≤0.1% (≤1,000ppm)
Appearance (Color)White
Yttrium(III) chloride hydrate acts as a source to prepare highly active and enantioselective yttrium precatalysts for asymmetric hydroamination of gem-disubstituted aminoalkenes. It is used as a precursor for the synthesis of yttrium oxide nanocrystallites. It employs in the crystal structure determination of macromolecules such as proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only