Thermo Scientific Chemicals

Yttrium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011185.22
100 g, Each
Thermo Scientific Chemicals

Yttrium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011185.22
100 g, Each
Quantity
Catalog number: 011185.22
also known as 011185-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12741-05-8
IUPAC Name
yttrium(3+) trichloride
Molecular Formula
Cl3Y
InChI Key
PCMOZDDGXKIOLL-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Y+3]
Total Rare Earth Oxide Impurities
0.01% max.

Description

Yttrium(III) chloride is used a precursor for the synthesis of optically active crystals and as a starting material for the synthesis of polymerization catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text