Yttrium(III) acetate tetrahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Yttrium(III) acetate tetrahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium(III) acetate tetrahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 011186.14
also known as 011186-14
Price (USD)/ Each
47.20
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
47.20
Add to cart
Yttrium(III) acetate tetrahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number011186.14
Price (USD)/ Each
47.20
-
Add to cart
Chemical Identifiers
CAS85949-60-6
IUPAC Nameyttrium(3+) triacetate tetrahydrate
Molecular FormulaC6H17O10Y
InChI KeyAIQRTHPXPDTMBQ-UHFFFAOYSA-K
SMILESO.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Yttrium(III) acetate tetrahydrate is used as laboratory reagents, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only