Thermo Scientific Chemicals

Yttrium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011187.14
25 g, Each
Thermo Scientific Chemicals

Yttrium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011187.14
25 g, Each
Quantity
Catalog number: 011187.14
also known as 011187-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
140191-40-8
IUPAC Name
yttrium(3+) trinitrate
Molecular Formula
N3O9Y
InChI Key
BXJPTTGFESFXJU-UHFFFAOYSA-N
SMILES
[Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Rare Earth Oxide Impurities
0.01% max.

Description

It acts as a precursor for the preparation of yttrium oxide nano particles and for nanoscale coatings of carbon composites. It is an effective yttrium source for the preparation of yttrium-based surfactant mesophases which are used as adsorbing agents and in optically functional materials. It is used in optics, ceramics, glass, electronics, and superconducting materials. It has been reported as a reusable catalyst in Bignelli reaction for the synthesis of heterocycles such as pyrimidin-2-ones. It is a powerful catalyst for the synthesis of some supramolecules such as calix[4]resorcinarenes, 1,8-dioxooctahydroxanthenes, 2-amino-4H-chromenes, and several other organic condensation products. Silica-supported yttrium nitrate hydrate acts as a heterogeneous catalyst for the preparation of 2-Amino-4H-Chromenes in water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text