Thermo Scientific Chemicals

Ytterbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011189.09
10 g, Each
Thermo Scientific Chemicals

Ytterbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011189.09
10 g, Each
Quantity
Catalog number: 011189.09
also known as 011189-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13760-80-0
IUPAC Name
ytterbium(3+) trifluoride
Molecular Formula
F3Yb
InChI Key
XASAPYQVQBKMIN-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Yb+3]
Appearance (Color)
White
Form
Powder
Total Rare Earth Oxide Impurities
≤0.02%

Description

Ytterbium(III) fluoride is used in the preparation and study of fluorinated glasses. Further, it is employed as a laboratory reagent and in optical glasses and structural ceramics. It finds application in catalysts, electrical components and photo-optical materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text