Ytterbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Ytterbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ytterbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 011193.09
also known as 011193-09
Price (USD)/ Each
48.65
Online exclusive
53.50 
Save 4.85 (9%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
48.65
Online exclusive
53.50 
Save 4.85 (9%)
Add to cart
Ytterbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Catalog number011193.09
Price (USD)/ Each
48.65
Online exclusive
53.50 
Save 4.85 (9%)
-
Add to cart
Chemical Identifiers
CAS19423-87-1
IUPAC Nameytterbium(3+) trichloride
Molecular FormulaCl3Yb
InChI KeyCKLHRQNQYIJFFX-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Yb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Ytterbium(III) chloride hydrate is used as an effective catalyst to prepare acetals by reacting with trimethyl orthoformate. It reacts with N-alkylsalicylideneimines to prepare yttrium(III) Schiff base complexes. It is widely used as NMR shift reagent, calcium ion probe and in DNA microarrays. Its anhydrous form acts as a Lewis acid and catalyzes the alkylation reactions such as aldol reaction and Pictet-Spengler reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only