Thermo Scientific Chemicals

Ytterbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011193.18
50 g, Each
Thermo Scientific Chemicals

Ytterbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011193.18
50 g, Each
Quantity
Catalog number: 011193.18
also known as 011193-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19423-87-1
IUPAC Name
ytterbium(3+) trichloride
Molecular Formula
Cl3Yb
InChI Key
CKLHRQNQYIJFFX-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Yb+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Ytterbium(III) chloride hydrate is used as an effective catalyst to prepare acetals by reacting with trimethyl orthoformate. It reacts with N-alkylsalicylideneimines to prepare yttrium(III) Schiff base complexes. It is widely used as NMR shift reagent, calcium ion probe and in DNA microarrays. Its anhydrous form acts as a Lewis acid and catalyzes the alkylation reactions such as aldol reaction and Pictet-Spengler reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text