Ytterbium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Ytterbium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ytterbium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Cl3H2OYb, CAS Number-19423-87-1, acmc-1bo6s, ytterbium 3+ chloride-water 1/3/1, ytterbium 3+ ion hydrate trichloride, ytterbium iii chloride hydrate, 50g, 99.99% (REO), White, 2.58, 233-800-5, 279.40 (Anhydrous), H315-H319-H335
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 011194.18
also known as 011194-18
Price (USD)/ Each
324.65
Online exclusive
361.00 
Save 36.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
324.65
Online exclusive
361.00 
Save 36.35 (10%)
Add to cart
Ytterbium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number011194.18
Price (USD)/ Each
324.65
Online exclusive
361.00 
Save 36.35 (10%)
-
Add to cart
Chemical Identifiers
CAS19423-87-1
IUPAC Nameytterbium(3+) trichloride
Molecular FormulaCl3Yb
InChI KeyCKLHRQNQYIJFFX-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Yb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Ytterbium(III) chloride hydrate is used as a catalyst in organic chemical synthesis. It is also used to track digestion in animals. The catalytic nature of this compound also has an application in DNA microarrays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only