Thermo Scientific Chemicals
Ytterbium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number: 011196.09
10 g, Each
Thermo Scientific Chemicals
Ytterbium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number: 011196.09
10 g, Each
Quantity
Catalog number: 011196.09
also known as 011196-09
Price (USD)
Quantity
-
Have Questions?
Total Rare Earth Oxide Impurities
0.01% max.
Description
Ytterbium(III) nitrate hydrate, is used as a precursor of nanoscale coatings of carbon composites. Source of yttrium used to make yttrium-based surfactant mesophases which are promising as adsorbing agents as well as for optically functional species.