Thulium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thulium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thulium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 011198.04
also known as 011198-04
Price (USD)/ Each
125.00
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
125.00
Add to cart
Thulium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Catalog number011198.04
Price (USD)/ Each
125.00
-
Add to cart
Chemical Identifiers
CAS12036-44-1
IUPAC Namedithulium(3+) trioxidandiide
Molecular FormulaO3Tm2
InChI KeyZIKATJAYWZUJPY-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Tm+3].[Tm+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total Rare Earth Oxide Impurities≤0.1% (1000 ppm)
FormPowder
Thulium oxide is an excellent thulium source which finds use in glass, optical and ceramic applications. Further, it is used in the manufacturing of portable X-ray transmission device. In addition to this, it finds use as a nuclear reactor control material. Nanostructured thulium oxide acts as an efficient biosensor in the field of medicinal chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only