Thermo Scientific Chemicals

Terbium(III,IV) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011207.18
50 g, Each
Thermo Scientific Chemicals

Terbium(III,IV) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011207.18
50 g, Each
Quantity
Catalog number: 011207.18
also known as 011207-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12037-01-3
IUPAC Name
tetraoxotetraterboxane
Molecular Formula
O7Tb4
InChI Key
UWZVBPNIJCKCMC-UHFFFAOYSA-N
SMILES
O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Terbium(III,IV) oxide acts as a precursor for the preparation of other terbium compounds. It is used as a redox catalyst in reactions involving oxygen. At high temperatures, it can liberate oxygen. It finds applications in the manufacture of glass, optical and ceramic. Nanoparticles of terbium oxide are used as analytical reagents for the determination of drugs in food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text