Thermo Scientific Chemicals

Terbium(III) chloride hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011210.18
50 g, Each
Thermo Scientific Chemicals

Terbium(III) chloride hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011210.18
50 g, Each
Quantity
Catalog number: 011210.18
also known as 011210-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13798-24-8
IUPAC Name
terbium(3+) hexahydrate trichloride
Molecular Formula
Cl3H12O6Tb
InChI Key
ULJUVCOAZNLCJZ-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3]
Total Rare Earth Oxide Impurities
0.01% max.

Description

Terbium(III) chloride hexahydrate is used in the primary chemical intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text