Scandium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Scandium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Scandium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number 011217.06
also known as 011217-06
Price (USD)/ Each
257.65
Online exclusive
286.00 
Save 28.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
257.65
Online exclusive
286.00 
Save 28.35 (10%)
Add to cart
Scandium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number011217.06
Price (USD)/ Each
257.65
Online exclusive
286.00 
Save 28.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12060-08-1
IUPAC Namediscandium(3+) trioxidandiide
Molecular FormulaO3Sc2
InChI KeyHJGMWXTVGKLUAQ-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Sc+3].[Sc+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total Rare Earth Oxide Impurities≤0.01% (100 ppm)
FormPowder
Scandium oxide is resistant to heat and thermal shock, hence finds use in high-temperature systems. Further, it is used in electronic ceramics and glass composition. It is used as a starting material for the preparation of other scandium compounds. In addition to this, it has vacuum deposition applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only