Thermo Scientific Chemicals

Scandium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011217.14
25 g, Each
Thermo Scientific Chemicals

Scandium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011217.14
25 g, Each
Quantity
Catalog number: 011217.14
also known as 011217-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12060-08-1
IUPAC Name
discandium(3+) trioxidandiide
Molecular Formula
O3Sc2
InChI Key
HJGMWXTVGKLUAQ-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Sc+3].[Sc+3]
Appearance (Color)
White
Total Rare Earth Oxide Impurities
≤0.01% (100 ppm)
Form
Powder

Description

Scandium oxide is resistant to heat and thermal shock, hence finds use in high-temperature systems. Further, it is used in electronic ceramics and glass composition. It is used as a starting material for the preparation of other scandium compounds. In addition to this, it has vacuum deposition applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text