Praseodymium(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Praseodymium(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Praseodymium(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 011240.14
also known as 011240-14
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
Add to cart
Praseodymium(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number011240.14
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS15878-77-0
IUPAC Namepraseodymium(3+) hexahydrate trinitrate
Molecular FormulaH12N3O15Pr
InChI KeyLXXCECZPOWZKLC-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Praseodymium(III) nitrate hexahydrate is used in electrical components, optical glasses and photo-optical materials. Also find use as catalysts and laboratory reagent. In addition, it is used in the solvothermal process for formation of doped lanthanide oxysulfide. It is a precursor for the electrodeposition of praseodymium oxide for electrochemical applications. It is a starting material for the preparation of ceramic pigments, metal-organic frameworks and mixed conductors. It finds utility in the synthesis of other praseodymium compounds and complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only