Neodymium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Neodymium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Neodymium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 011245.14
also known as 011245-14
Price (USD)/ Each
160.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
160.00
Add to cart
Neodymium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number011245.14
Price (USD)/ Each
160.00
-
Add to cart
Chemical Identifiers
CAS13746-96-8
IUPAC Nameneodymium(3+) trinitrate
Molecular FormulaN3NdO9
InChI KeyCFYGEIAZMVFFDE-UHFFFAOYSA-N
SMILES[Nd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
It is used as pharmaceutical intermediates. It is used in the selective PVC membrane sensors. The sensor with ionophore (L1) exhibits significantly enhanced selectivity towards neodymium (III).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only