Neodymium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Neodymium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Neodymium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Catalog number 011250.14
also known as 011250-14
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
Add to cart
Neodymium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number011250.14
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1313-97-9
IUPAC Namedineodymium(3+) trioxidandiide
Molecular FormulaNd2O3
InChI KeyPLDDOISOJJCEMH-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Nd+3].[Nd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Neodymium(III) oxide is used as a catalyst in polymer chemistry. Glasses doped with neodymium(III) oxide are dichroic in nature and is used in welding goggles and other colored glasses. It finds use in solid-state lasers, carbon arc-light electrodes and enamels. It serves as the starting material for the synthesis of other neodymium(III) compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only