Thermo Scientific Chemicals

Neodymium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011250.A1
1 kg, Each
Thermo Scientific Chemicals

Neodymium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011250.A1
1 kg, Each
Quantity
Catalog number: 011250.A1
also known as 011250-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1313-97-9
IUPAC Name
dineodymium(3+) trioxidandiide
Molecular Formula
Nd2O3
InChI Key
PLDDOISOJJCEMH-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Nd+3].[Nd+3]
Total Rare Earth Oxide Impurities
0.01% max.

Description

Neodymium(III) oxide is used as a catalyst in polymer chemistry. Glasses doped with neodymium(III) oxide are dichroic in nature and is used in welding goggles and other colored glasses. It finds use in solid-state lasers, carbon arc-light electrodes and enamels. It serves as the starting material for the synthesis of other neodymium(III) compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text