Thermo Scientific Chemicals

Neodymium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011251.18
50 g, Each
Thermo Scientific Chemicals

Neodymium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011251.18
50 g, Each
Quantity
Catalog number: 011251.18
also known as 011251-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19423-78-0
IUPAC Name
neodymium(3+) trichloride
Molecular Formula
Cl3Nd
InChI Key
ATINCSYRHURBSP-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Nd+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Neodymium(III) chloride hydrate is used for production of neodymium metal. It also finds application in lasers and fiber amplifiers. It serves as dopants for silica based as well as plastic fibers. It plays an important role of additives in organic light emitting diodes resulting in improved LED emission spectrum. In addition, it is used in catalysis of polymerization of various dienes. It is used to modify titanium dioxide for use as a photocatalyst. Its coating on aluminum and its alloy protect them from corrosion. It is also extensively used as fluorescent labels for organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text