Thermo Scientific Chemicals

Lutetium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011255.03
1 g, Each
Thermo Scientific Chemicals

Lutetium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011255.03
1 g, Each
Quantity
Catalog number: 011255.03
also known as 011255-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12032-20-1
IUPAC Name
dilutetium(3+) trioxidandiide
Molecular Formula
Lu2O3
InChI Key
UGBIHFMRUDAMBY-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Lu+3].[Lu+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Lutetium(III) oxide serves as a catalyst in a variety of reactions such as polymerization, hydrogenation and alkylation reactions. It is also used in ceramics and glass. In the production of laser crystals, lutetium(III) oxide finds application as an important starting material. Its luminescent properties, a field of interest for researchers and may find potential application in biological fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text