Lutetium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Lutetium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lutetium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number 011256.06
also known as 011256-06
Price (USD)/ Each
320.65
Online exclusive
356.00 
Save 35.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
320.65
Online exclusive
356.00 
Save 35.35 (10%)
Add to cart
Lutetium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number011256.06
Price (USD)/ Each
320.65
Online exclusive
356.00 
Save 35.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12032-20-1
IUPAC Namedilutetium(3+) trioxidandiide
Molecular FormulaLu2O3
InChI KeyUGBIHFMRUDAMBY-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Lu+3].[Lu+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Lutetium(III) oxide serves as a catalyst in a variety of reactions such as polymerization, hydrogenation and alkylation reactions. It is also used in ceramics and glass. In the production of laser crystals, lutetium(III) oxide finds application as an important starting material. Its luminescent properties, a field of interest for researchers and may find potential application in biological fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only