Lutetium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Lutetium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lutetium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number 011258.09
also known as 011258-09
Price (USD)/ Each
161.65
Online exclusive
190.00 
Save 28.35 (15%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
161.65
Online exclusive
190.00 
Save 28.35 (15%)
Add to cart
Lutetium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number011258.09
Price (USD)/ Each
161.65
Online exclusive
190.00 
Save 28.35 (15%)
-
Add to cart
Chemical Identifiers
CAS100641-16-5
IUPAC Namelutetium(3+) trinitrate
Molecular FormulaLuN3O9
InChI KeyAPRNQTOXCXOSHO-UHFFFAOYSA-N
SMILES[Lu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only