Thermo Scientific Chemicals

Lutetium(III) chloride hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011260.06
5 g, Each
Thermo Scientific Chemicals

Lutetium(III) chloride hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011260.06
5 g, Each
Quantity
Catalog number: 011260.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15230-79-2
IUPAC Name
lutetium(3+) tris(nitric acid)
Molecular Formula
H3LuN3O9
InChI Key
MESQZVHAWZZUAY-UHFFFAOYSA-N
SMILES
[Lu+3].O[N+]([O-])=O.O[N+]([O-])=O.O[N+]([O-])=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Lutetium(III) chloride hexahydrate is used as a catalyst and laboratory reagent. It is also used in ceramics, optical glasses, electrical components and photo-optical material. It is one of the components of scintillating composition. It serves as a precursor for the preparation of other lutetium complexes. It finds application as one of the starting materials for aluminate phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text