Lanthanum(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Lanthanum(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number 011264.30
also known as 011264-30
Price (USD)/ Each
215.65
Online exclusive
239.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
215.65
Online exclusive
239.00 
Save 23.35 (10%)
Add to cart
Lanthanum(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number011264.30
Price (USD)/ Each
215.65
Online exclusive
239.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1312-81-8
IUPAC Namedilanthanum(3+) trioxidandiide
Molecular FormulaLa2O3
InChI KeyMRELNEQAGSRDBK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[La+3].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacturing of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics. In addition, it is used as a catalyst to effect certain organic transformations. It is one of the active ingredients and finds application in thermoelectric and piezoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only