Thermo Scientific Chemicals

Lanthanum(III) oxide, 99.99% (REO), Ca 10ppm max, AAS Grade, Thermo Scientific Chemicals

Catalog number: 011266.30
250 g, Each
Thermo Scientific Chemicals

Lanthanum(III) oxide, 99.99% (REO), Ca 10ppm max, AAS Grade, Thermo Scientific Chemicals

Catalog number: 011266.30
250 g, Each
Quantity
Catalog number: 011266.30
also known as 011266-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1312-81-8
IUPAC Name
dilanthanum(3+) trioxidandiide
Molecular Formula
La2O3
InChI Key
MRELNEQAGSRDBK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[La+3].[La+3]
Appearance (Color)
White
Impurity content
Ca ≤ 10ppm
Total Rare Earth Oxide Impurities
≤0.01% (100 ppm)
Form
Powder

Description

Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacturing of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics. In addition, it is used as a catalyst to effect certain organic transformations. It is one of the active ingredients and finds application in thermoelectric and piezoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text