Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
1 kg
Catalog number 011267.06
also known as 011267-06
Price (USD)/ Each
38.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
38.00
Add to cart
Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number011267.06
Price (USD)/ Each
38.00
-
Add to cart
Chemical Identifiers
CAS10277-43-7
IUPAC Namelanthanum(3+) hexahydrate trinitrate
Molecular FormulaH12LaN3O15
InChI KeyGJKFIJKSBFYMQK-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Lanthanum(III) nitrate is utilized as a starting material for the electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. It is used as a catalyst for the chemoselective preparation of cyclic and acyclic dithioacetals. It is used to prepare lanthanum aluminate (LAO) thin films, LaF3 nanocrystals, and bis(indolyl) methanes from indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only