Thermo Scientific Chemicals

Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011267.30
250 g, Each
Thermo Scientific Chemicals

Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011267.30
250 g, Each
Quantity
Catalog number: 011267.30
also known as 011267-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10277-43-7
IUPAC Name
lanthanum(3+) hexahydrate trinitrate
Molecular Formula
H12LaN3O15
InChI Key
GJKFIJKSBFYMQK-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Rare Earth Oxide Impurities
0.01% max.

Description

Lanthanum(III) nitrate is utilized as a starting material for the electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. It is used as a catalyst for the chemoselective preparation of cyclic and acyclic dithioacetals. It is used to prepare lanthanum aluminate (LAO) thin films, LaF3 nanocrystals, and bis(indolyl) methanes from indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text