Thermo Scientific Chemicals

Lanthanum nickel, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011271.90
5 x 50 g, Each
Thermo Scientific Chemicals

Lanthanum nickel, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011271.90
5 x 50 g, Each
Quantity
Catalog number: 011271.90
also known as 011271-90
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12196-72-4
IUPAC Name
lanthanum pentanickel
Molecular Formula
LaNi5
InChI Key
BUCVQDCWULHLJZ-UHFFFAOYSA-N
SMILES
[Ni].[Ni].[Ni].[Ni].[Ni].[La]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Lanthanum nickel act as a superconducting intermetallic phase in lanthanum nickel boro-nitride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text