Thermo Scientific Chemicals

Lanthanum(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011272.36
500 g, Each
Thermo Scientific Chemicals

Lanthanum(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011272.36
500 g, Each
Quantity
Catalog number: 011272.36
also known as 011272-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1312-81-8
IUPAC Name
dilanthanum(3+) trioxidandiide
Molecular Formula
La2O3
InChI Key
MRELNEQAGSRDBK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[La+3].[La+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacture of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text