Holmium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Holmium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Holmium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 011280.09
also known as 011280-09
Price (USD)/ Each
79.65
Online exclusive
88.50 
Save 8.85 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
79.65
Online exclusive
88.50 
Save 8.85 (10%)
Add to cart
Holmium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Catalog number011280.09
Price (USD)/ Each
79.65
Online exclusive
88.50 
Save 8.85 (10%)
-
Add to cart
Chemical Identifiers
CAS12055-62-8
IUPAC Namediholmium(3+) trioxidandiide
Molecular FormulaHo2O3
InChI KeyOWCYYNSBGXMRQN-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Ho+3].[Ho+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Holmium(III) oxide iacts as colorants which is used for cubic zirconia and glass, as a calibration standard for optical spectrophotometers, as a specialty catalyst, phosphor and a laser material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only