Thermo Scientific Chemicals

Gadolinium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011284.14
25 g, Each
Thermo Scientific Chemicals

Gadolinium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011284.14
25 g, Each
Quantity
Catalog number: 011284.14
also known as 011284-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
94219-55-3
IUPAC Name
gadolinium(3+) trinitrate
Molecular Formula
GdN3O9
InChI Key
MWFSXYMZCVAQCC-UHFFFAOYSA-N
SMILES
[Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Rare Earth Oxide Impurities
0.01% max.

Description

Gadolinium(III) nitrate is used as an oxidizing agent and the precursor material in the preparation of gadolinium compounds. It is extensively employed in the production of specialized glasses and ceramics. It is also employed in heavy water nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text