Thermo Scientific Chemicals

Gadolinium(III) chloride hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011287.22
100 g, Each
Thermo Scientific Chemicals

Gadolinium(III) chloride hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011287.22
100 g, Each
Quantity
Catalog number: 011287.22
also known as 011287-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13450-84-5
IUPAC Name
gadolinium(3+) hexahydrate trichloride
Molecular Formula
Cl3GdH12O6
InChI Key
PNYPSKHTTCTAMD-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Gd+3]
Total Rare Earth Oxide Impurities
0.01% max.

Description

Gadolinium(III) chloride hexahydrate is employed in synthesis and NMR studies of the paramagnetic GdPCTA-[12] and GdPCTA-[13] complexes with possible use as contrast agents for magnetic resonance imaging..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text