Thermo Scientific Chemicals

Erbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011304.14
25 g, Each
Thermo Scientific Chemicals

Erbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011304.14
25 g, Each
Quantity
Catalog number: 011304.14
also known as 011304-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19423-85-9
IUPAC Name
erbium(3+) trichloride
Molecular Formula
Cl3Er
InChI Key
HDGGAKOVUDZYES-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Er+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Erbium(III) chloride hydrate is used as a starting material for new edge-bridged octahedral M6 cluster compounds such as CsErTa6Cl18, for which electron and stability studies were performed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text