Thermo Scientific Chemicals

Erbium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011305.14
25 g, Each
Thermo Scientific Chemicals

Erbium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011305.14
25 g, Each
Quantity
Catalog number: 011305.14
also known as 011305-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19423-85-9
IUPAC Name
erbium(3+) trichloride
Molecular Formula
Cl3Er
InChI Key
HDGGAKOVUDZYES-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Er+3]
Total Rare Earth Oxide Impurities
0.01% max.

Description

Erbium(III) chloride hydrate serves as a mild Lewis acid for selective acetalization. It combines with sodium borohydride and used as a selective reducing agent. It is also useful to form enolates from alfa-bromo ketones and oxyallyl cations from alfa,alfa'-dibromo ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text