Erbium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Erbium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Erbium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 011306.14
also known as 011306-14
Price (USD)/ Each
161.65
Online exclusive
179.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
161.65
Online exclusive
179.00 
Save 17.35 (10%)
Add to cart
Erbium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number011306.14
Price (USD)/ Each
161.65
Online exclusive
179.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10031-51-3
IUPAC Nameerbium(3+) pentahydrate trinitrate
Molecular FormulaErH10N3O14
InChI KeyLWHHUEHWVBVASY-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Erbium(III) nitrate pentahydrate is used as a chemical intermediates, producing a catalyst and related compounds, and also primary and secondary intermediated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only