Thermo Scientific Chemicals

Erbium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011306.22
100 g, Each
Thermo Scientific Chemicals

Erbium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011306.22
100 g, Each
Quantity
Catalog number: 011306.22
also known as 011306-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10031-51-3
IUPAC Name
erbium(3+) pentahydrate trinitrate
Molecular Formula
ErH10N3O14
InChI Key
LWHHUEHWVBVASY-UHFFFAOYSA-N
SMILES
O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Rare Earth Oxide Impurities
0.01% max.

Description

Erbium(III) nitrate pentahydrate is used as a chemical intermediates, producing a catalyst and related compounds, and also primary and secondary intermediated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text