Erbium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Erbium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Erbium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
500 g
Catalog number 011309.22
also known as 011309-22
Price (USD)/ Each
531.65
Online exclusive
591.00 
Save 59.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
531.65
Online exclusive
591.00 
Save 59.35 (10%)
Add to cart
Erbium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number011309.22
Price (USD)/ Each
531.65
Online exclusive
591.00 
Save 59.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12061-16-4
IUPAC Namedierbium(3+) trioxidandiide
Molecular FormulaEr2O3
InChI KeyZXGIFJXRQHZCGJ-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Er+3].[Er+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Erbium trioxide is an important rare earth metal used in biomedicine. It is used as a gate dielectric in semi conductor devices since it has a high dielectric constant and a large band gap; as a burnable neutron poison for nuclear fuel and as coloring for glasses. Erbium oxide nanoparticles can be surface modified for distribution into aqueous and non-aqueous media for bioimaging due to its photoluminescence property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only