Erbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Erbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Erbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 011312.18
also known as 011312-18
Price (USD)/ Each
423.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
423.00
Add to cart
Erbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number011312.18
Price (USD)/ Each
423.00
-
Add to cart
Chemical Identifiers
CAS13760-83-3
IUPAC Nameerbium(3+) trifluoride
Molecular FormulaErF3
InChI KeyQGJSAGBHFTXOTM-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Er+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Erbium(III) fluoride is an important colorant in glass manufacturing and porcelain enamel glazes. It is used to produce erbium metal. It is used as dopant in making optical products. It is highly employed as erbium doped fluoride fibers in optical communications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only