Thermo Scientific Chemicals

Dysprosium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011315.22
100 g, Each
Thermo Scientific Chemicals

Dysprosium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011315.22
100 g, Each
Quantity
Catalog number: 011315.22
also known as 011315-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10031-49-9
IUPAC Name
dysprosium(3+) pentahydrate trinitrate
Molecular Formula
DyH10N3O14
InChI Key
NOTQUFQJAWMLCE-UHFFFAOYSA-N
SMILES
O.O.O.O.O.[Dy+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Rare Earth Oxide Impurities
0.01% max.

Description

Dysprosium(III) nitrate is used for catalysts. It is used in the preparation of dysprosium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text