Dysprosium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Dysprosium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dysprosium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

For Use in Cermets Used As Nuclear Reaction Control Rods
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 011319.09
also known as 011319-09
Price (USD)/ Each
56.65
Online exclusive
63.10 
Save 6.45 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
56.65
Online exclusive
63.10 
Save 6.45 (10%)
Add to cart
Dysprosium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Catalog number011319.09
Price (USD)/ Each
56.65
Online exclusive
63.10 
Save 6.45 (10%)
-
Add to cart
Chemical Identifiers
CAS1308-87-8
IUPAC Namedidysprosium(3+) trioxidandiide
Molecular FormulaDy2O3
InChI KeyGEZAXHSNIQTPMM-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Dy+3].[Dy+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
In cermets used as nuclear reaction control rodsDysprosium trioxide is used with nickel in cermets, which are used as nuclear reactor control rods. It is also used in ceramics, glass, phosphors, lasers and dysprosium metal halide lamps. It is an important raw material to prepare dysprosium(III) salts by reacting with acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only