Cerium(III) nitrate hexahydrate, REacton™, 99.5% (REO), Thermo Scientific Chemicals
Cerium(III) nitrate hexahydrate, REacton™, 99.5% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cerium(III) nitrate hexahydrate, REacton™, 99.5% (REO), Thermo Scientific Chemicals

For Separation of Cerium From Other Rare Earths, Catalyst For Hydrolysis of Phosporic Acid Esters
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
100 g
1 kg
Catalog number 011329.36
also known as 011329-36
Price (USD)/ Each
312.65
Online exclusive
347.00 
Save 34.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
312.65
Online exclusive
347.00 
Save 34.35 (10%)
Add to cart
Cerium(III) nitrate hexahydrate, REacton™, 99.5% (REO), Thermo Scientific Chemicals
Catalog number011329.36
Price (USD)/ Each
312.65
Online exclusive
347.00 
Save 34.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10294-41-4
IUPAC Namecerium(3+) hexahydrate trinitrate
Molecular FormulaCeH12N3O15
InChI KeyQQZMWMKOWKGPQY-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥99.5% (CeO2/TREO)
Total Rare Earth Oxide Impurities≤0.5%
Cerium(III) nitrate hexahydrate is used in the separation of cerium from other rare earths. It plays an important role as a catalyst for hydrolysis of phosphoric acid esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only