Thermo Scientific Chemicals

Cerium(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011330.06
5 g, Each
Thermo Scientific Chemicals

Cerium(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011330.06
5 g, Each
Quantity
Catalog number: 011330.06
also known as 011330-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10294-41-4
IUPAC Name
cerium(3+) hexahydrate trinitrate
Molecular Formula
CeH12N3O15
InChI Key
QQZMWMKOWKGPQY-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Form
Crystalline aggregates, crystals or crystalline powder
Assay (special)
38.8-40.5% (CeO2)
Comment
Specification differs for U.S. and non-U.S. material where indicated
Impurity content
Calcium oxide (CaO): ≤25 ppm (non-U.S. specification)
Impurity content
Iron (III) oxide (Fe2O3): ≤25 ppm (non-U.S. specification)

Description

Cerium(III) nitrate hexahydrate is used in the separation of cerium from other rare earths. It plays an important role as a catalyst for hydrolysis of phosphoric acid esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text